Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613082 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP2C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ATP2C2 - ATPase secretory pathway Ca2+ transporting 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286527.2 | Intron | NP_001273456.2 | ||||
NM_001291454.1 | Intron | NP_001278383.1 | ||||
NM_014861.3 | Intron | NP_055676.3 | ||||
XM_011523486.2 | Intron | XP_011521788.1 | ||||
XM_011523487.2 | Intron | XP_011521789.1 |
Set Membership: |
HapMap |