Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603333 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAH2 - dynein axonemal heavy chain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303270.1 | Intron | NP_001290199.1 | ||||
NM_020877.3 | Intron | NP_065928.2 | ||||
XM_011523663.1 | Intron | XP_011521965.1 | ||||
XM_011523664.2 | Intron | XP_011521966.1 | ||||
XM_011523666.1 | Intron | XP_011521968.1 | ||||
XM_011523667.2 | Intron | XP_011521969.1 | ||||
XM_011523668.1 | Intron | XP_011521970.1 | ||||
XM_011523669.2 | Intron | XP_011521971.1 | ||||
XM_011523670.2 | Intron | XP_011521972.1 | ||||
XM_017024218.1 | Intron | XP_016879707.1 | ||||
XM_017024219.1 | Intron | XP_016879708.1 |