Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604601 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRF1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTRF1 - mitochondrial translational release factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAA16 - N(alpha)-acetyltransferase 16, NatA auxiliary subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110798.1 | Intron | NP_001104268.1 | ||||
NM_018527.3 | Intron | NP_060997.2 | ||||
NM_024561.4 | Intron | NP_078837.3 | ||||
XM_005266523.2 | Intron | XP_005266580.1 | ||||
XM_006719866.2 | Intron | XP_006719929.1 | ||||
XM_011535227.2 | Intron | XP_011533529.1 | ||||
XM_011535228.1 | Intron | XP_011533530.1 | ||||
XM_017020744.1 | Intron | XP_016876233.1 | ||||
XM_017020745.1 | Intron | XP_016876234.1 | ||||
XM_017020746.1 | Intron | XP_016876235.1 |
Set Membership: |
HapMap |