Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPACA7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SPACA7 - sperm acrosome associated 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145248.4 | Intron | NP_660291.2 | ||||
XM_011537466.1 | Intron | XP_011535768.1 | ||||
XM_011537467.2 | Intron | XP_011535769.1 | ||||
XM_011537468.2 | Intron | XP_011535770.1 | ||||
XM_011537471.2 | Intron | XP_011535773.1 | ||||
XM_011537472.2 | Intron | XP_011535774.1 | ||||
XM_017020391.1 | Intron | XP_016875880.1 | ||||
XM_017020392.1 | Intron | XP_016875881.1 |