Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCATCTGTATCACCCCTAGTAGAC[A/G]CGAGGGTTTCCCCAATTACATGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616598 MIM: 607175 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BORCS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BORCS5 - BLOC-1 related complex subunit 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUSP16 - dual specificity phosphatase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030640.2 | 4846 | UTR 3 | NP_085143.1 | |||
XM_005253488.2 | 4846 | UTR 3 | XP_005253545.1 | |||
XM_006719155.2 | 4846 | UTR 3 | XP_006719218.1 | |||
XM_011520856.1 | 4846 | UTR 3 | XP_011519158.1 | |||
XM_011520857.1 | 4846 | UTR 3 | XP_011519159.1 | |||
XM_017019988.1 | 4846 | UTR 3 | XP_016875477.1 |
LOC105369662 - uncharacterized LOC105369662 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |