Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176806 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTGER3 - prostaglandin E receptor 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126044.1 | Intron | NP_001119516.1 | ||||
NM_198714.1 | Intron | NP_942007.1 | ||||
NM_198715.2 | Intron | NP_942008.1 | ||||
NM_198716.1 | Intron | NP_942009.1 | ||||
NM_198717.1 | Intron | NP_942010.1 | ||||
NM_198718.1 | Intron | NP_942011.1 | ||||
NM_198719.1 | Intron | NP_942012.1 | ||||
XM_011541810.2 | Intron | XP_011540112.1 |
Set Membership: |
HapMap |