Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD207 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD207 - CD207 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015717.4 | Intron | NP_056532.4 | ||||
XM_011532874.1 | Intron | XP_011531176.1 | ||||
XM_011532875.2 | Intron | XP_011531177.1 | ||||
XM_011532876.2 | Intron | XP_011531178.1 |
CLEC4F - C-type lectin domain family 4 member F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |