Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614694 MIM: 614425 | ||||||||||||||||||||||||||||||||
Literature Links: |
RPRD1B PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
RPRD1B - regulation of nuclear pre-mRNA domain containing 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021215.3 | Intron | NP_067038.1 | ||||
XM_005260477.3 | Intron | XP_005260534.1 | ||||
XM_005260480.4 | Intron | XP_005260537.1 | ||||
XM_011528954.2 | Intron | XP_011527256.1 | ||||
XM_017027990.1 | Intron | XP_016883479.1 | ||||
XM_017027991.1 | Intron | XP_016883480.1 |
TTI1 - TELO2 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303457.1 | Intron | NP_001290386.1 | ||||
NM_014657.2 | Intron | NP_055472.1 | ||||
XM_011529114.1 | Intron | XP_011527416.1 | ||||
XM_017028148.1 | Intron | XP_016883637.1 |
Set Membership: |
HapMap |