Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCGGAGGCAATCACATTTATCCC[C/T]TTTTGTTTTTATTTCTTTGGATATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 150340 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMNB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMNB1 - lamin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198557.1 | Intron | NP_001185486.1 | ||||
NM_005573.3 | Intron | NP_005564.1 |
LOC102723557 - uncharacterized LOC102723557 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |