Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604778 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM29 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM29 - ADAM metallopeptidase domain 29 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130703.1 | 160 | UTR 5 | NP_001124175.1 | |||
NM_001130704.1 | 160 | Intron | NP_001124176.1 | |||
NM_001130705.1 | 160 | Intron | NP_001124177.1 | |||
NM_001278125.1 | 160 | UTR 5 | NP_001265054.1 | |||
NM_001278126.1 | 160 | UTR 5 | NP_001265055.1 | |||
NM_001278127.1 | 160 | Intron | NP_001265056.1 | |||
NM_014269.4 | 160 | UTR 5 | NP_055084.3 | |||
XM_011531556.1 | 160 | UTR 5 | XP_011529858.1 | |||
XM_011531557.1 | 160 | UTR 5 | XP_011529859.1 | |||
XM_011531559.1 | 160 | UTR 5 | XP_011529861.1 | |||
XM_011531560.1 | 160 | UTR 5 | XP_011529862.1 | |||
XM_011531561.1 | 160 | UTR 5 | XP_011529863.1 |