Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616695 MIM: 616550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
STYXL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
STYXL1 - serine/threonine/tyrosine interacting-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317785.1 | Intron | NP_001304714.1 | ||||
NM_001317786.1 | Intron | NP_001304715.1 | ||||
NM_001317787.1 | Intron | NP_001304716.1 | ||||
NM_001317788.1 | Intron | NP_001304717.1 | ||||
NM_001317789.1 | Intron | NP_001304718.1 | ||||
NM_016086.2 | Intron | NP_057170.1 | ||||
XM_011516292.2 | Intron | XP_011514594.1 | ||||
XM_011516293.2 | Intron | XP_011514595.1 | ||||
XM_011516300.1 | Intron | XP_011514602.1 | ||||
XM_017012296.1 | Intron | XP_016867785.1 | ||||
XM_017012297.1 | Intron | XP_016867786.1 | ||||
XM_017012298.1 | Intron | XP_016867787.1 | ||||
XM_017012299.1 | Intron | XP_016867788.1 | ||||
XM_017012300.1 | Intron | XP_016867789.1 | ||||
XM_017012301.1 | Intron | XP_016867790.1 | ||||
XM_017012302.1 | Intron | XP_016867791.1 |
TMEM120A - transmembrane protein 120A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |