Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 124015 MIM: 616695 MIM: 616550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POR - cytochrome p450 oxidoreductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STYXL1 - serine/threonine/tyrosine interacting-like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM120A - transmembrane protein 120A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317803.1 | Intron | NP_001304732.1 | ||||
NM_031925.2 | Intron | NP_114131.1 |