Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608119 | ||||||||||||||||||||
Literature Links: |
HOMEZ PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HOMEZ - homeobox and leucine zipper encoding | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020834.2 | 2064 | UTR 3 | NP_065885.2 |
RNF212B - ring finger protein 212B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282322.1 | 2064 | UTR 3 | NP_001269251.1 | |||
XM_011536313.2 | 2064 | Intron | XP_011534615.1 | |||
XM_011536314.2 | 2064 | Intron | XP_011534616.1 | |||
XM_011536315.2 | 2064 | Intron | XP_011534617.1 | |||
XM_011536316.2 | 2064 | Intron | XP_011534618.1 | |||
XM_011536319.2 | 2064 | Intron | XP_011534621.1 | |||
XM_011536322.2 | 2064 | Intron | XP_011534624.1 | |||
XM_011536323.2 | 2064 | Intron | XP_011534625.1 | |||
XM_011536324.2 | 2064 | Intron | XP_011534626.1 | |||
XM_011536327.2 | 2064 | Intron | XP_011534629.1 | |||
XM_011536328.2 | 2064 | Intron | XP_011534630.1 | |||
XM_011536334.2 | 2064 | Intron | XP_011534636.1 |