Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTTCCTTGGGGCTCCTCTGGC[C/T]GAGGAGCTGCTGGAAGAGGTTCCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM186B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM186B - family with sequence similarity 186 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032130.2 | Intron | NP_115506.1 | ||||
XM_006719625.2 | Intron | XP_006719688.1 | ||||
XM_006719626.2 | Intron | XP_006719689.1 | ||||
XM_006719627.3 | Intron | XP_006719690.1 | ||||
XM_011538796.2 | Intron | XP_011537098.1 | ||||
XM_011538797.2 | Intron | XP_011537099.1 | ||||
XM_017020008.1 | Intron | XP_016875497.1 |
LOC100422296 - Parkinsonism associated deglycase pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |