Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608170 MIM: 611435 MIM: 615813 | ||||||||||||||||||||||||||||||||
Literature Links: |
DDX41 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
DDX41 - DEAD-box helicase 41 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DOK3 - docking protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM193B - family with sequence similarity 193 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190946.2 | 3378 | UTR 3 | NP_001177875.1 | |||
XM_006714875.2 | 3378 | UTR 3 | XP_006714938.1 | |||
XM_006714876.1 | 3378 | UTR 3 | XP_006714939.1 | |||
XM_006714878.1 | 3378 | UTR 3 | XP_006714941.1 | |||
XM_006714879.3 | 3378 | UTR 3 | XP_006714942.1 | |||
XM_006714880.1 | 3378 | UTR 3 | XP_006714943.1 | |||
XM_006714884.1 | 3378 | UTR 3 | XP_006714947.1 | |||
XM_006714885.1 | 3378 | UTR 3 | XP_006714948.1 | |||
XM_006714886.2 | 3378 | Intron | XP_006714949.1 | |||
XM_011534572.1 | 3378 | UTR 3 | XP_011532874.1 | |||
XM_011534573.1 | 3378 | UTR 3 | XP_011532875.1 | |||
XM_011534574.1 | 3378 | UTR 3 | XP_011532876.1 | |||
XM_011534579.2 | 3378 | UTR 3 | XP_011532881.1 | |||
XM_011534580.1 | 3378 | UTR 3 | XP_011532882.1 | |||
XM_011534581.1 | 3378 | UTR 3 | XP_011532883.1 | |||
XM_011534582.1 | 3378 | UTR 3 | XP_011532884.1 | |||
XM_011534583.1 | 3378 | UTR 3 | XP_011532885.1 |