Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGCCAGCCAACTTGCTCTTCC[C/T]TGCGCTGTAGCCAGAGACCGGGCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
OR13A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
OR13A1 - olfactory receptor family 13 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004297.2 | 4941 | Missense Mutation | AGG,GGG | R,G 287 | NP_001004297.2 | |
XM_011540147.2 | 4941 | Missense Mutation | AGG,GGG | R,G 287 | XP_011538449.1 | |
XM_017016632.1 | 4941 | Missense Mutation | AGG,GGG | R,G 287 | XP_016872121.1 | |
XM_017016633.1 | 4941 | Missense Mutation | AGG,GGG | R,G 287 | XP_016872122.1 | |
XM_017016634.1 | 4941 | Missense Mutation | AGG,GGG | R,G 287 | XP_016872123.1 |