Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCGGCCACCGACAGTCGCCACCC[A/G]CGCTCCCCAGACCCTTAACCCCTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FOXI2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXI2 - forkhead box I2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207426.2 | 845 | Missense Mutation | CAC,CGC | H,R 282 | NP_997309.2 |
LOC387720 - collagen alpha-1(X) chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |