Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACAGTAGTCACAGGCGAAGGGC[C/T]TCTCACCGGTGTGGGTGCGGATATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611392 MIM: 129010 | ||||||||||||||||||||
Literature Links: |
ADO PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADO - 2-aminoethanethiol dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
EGR2 - early growth response 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000399.4 | 1396 | Missense Mutation | AAG,AGG | K,R 396 | NP_000390.2 | |
NM_001136177.2 | 1396 | Missense Mutation | AAG,AGG | K,R 396 | NP_001129649.1 | |
NM_001136178.1 | 1396 | Missense Mutation | AAG,AGG | K,R 396 | NP_001129650.1 | |
NM_001136179.2 | 1396 | Missense Mutation | AAG,AGG | K,R 346 | NP_001129651.1 | |
NM_001321037.1 | 1396 | Missense Mutation | AAG,AGG | K,R 346 | NP_001307966.1 | |
XM_011539427.1 | 1396 | Missense Mutation | AAG,AGG | K,R 409 | XP_011537729.1 |