Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAACGGAATGCAAGGAGATTGCA[A/G]TCTGGTTAGTGAAATAGAATGGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608734 | ||||||||||||||||||||
Literature Links: |
SLC39A12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC39A12 - solute carrier family 39 member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145195.1 | 530 | Missense Mutation | AAT,AGT | N,S 86 | NP_001138667.1 | |
NM_001282733.1 | 530 | Missense Mutation | AAT,AGT | N,S 86 | NP_001269662.1 | |
NM_001282734.1 | 530 | Intron | NP_001269663.1 | |||
NM_152725.3 | 530 | Missense Mutation | AAT,AGT | N,S 86 | NP_689938.2 |