Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCATCCTGCAGCTTCAGCGTGAGC[A/G]TGTCATAGTTGGCTCTCACCACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604661 | ||||||||||||||||||||
Literature Links: |
C10orf76 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C10orf76 - chromosome 10 open reading frame 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024541.2 | 2039 | Missense Mutation | ACG,ATG | T,M 631 | NP_078817.2 | |
XM_006717969.1 | 2039 | Missense Mutation | ACG,ATG | T,M 561 | XP_006718032.1 | |
XM_011540151.2 | 2039 | Missense Mutation | ACG,ATG | T,M 631 | XP_011538453.1 | |
XM_011540152.2 | 2039 | Missense Mutation | ACG,ATG | T,M 609 | XP_011538454.1 | |
XM_011540153.2 | 2039 | Missense Mutation | ACG,ATG | T,M 597 | XP_011538455.1 | |
XM_011540154.2 | 2039 | Missense Mutation | ACG,ATG | T,M 575 | XP_011538456.1 | |
XM_011540156.2 | 2039 | Missense Mutation | ACG,ATG | T,M 457 | XP_011538458.1 | |
XM_011540157.2 | 2039 | Missense Mutation | ACG,ATG | T,M 580 | XP_011538459.1 | |
XM_017016635.1 | 2039 | UTR 3 | XP_016872124.1 | |||
XM_017016636.1 | 2039 | Intron | XP_016872125.1 |
KCNIP2 - potassium voltage-gated channel interacting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |