Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGGGCTTTTCTTGTCTTCTCC[C/T]TGGTCATCATCATCATCGTCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM171A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM171A1 - family with sequence similarity 171 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010924.1 | 2532 | Silent Mutation | CAA,CAG | Q,Q 868 | NP_001010924.1 | |
XM_011519378.2 | 2532 | Silent Mutation | CAA,CAG | Q,Q 855 | XP_011517680.1 | |
XM_017015904.1 | 2532 | Silent Mutation | CAA,CAG | Q,Q 839 | XP_016871393.1 | |
XM_017015905.1 | 2532 | Silent Mutation | CAA,CAG | Q,Q 510 | XP_016871394.1 |