Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCTTGCCCACCTTCTCCCAGCC[C/T]TTCGGGGGTGGCTCCCACATGGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164012 MIM: 602327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFKB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFKB2 - nuclear factor kappa B subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077494.3 | 1320 | Silent Mutation | CCC,CCT | P,P 349 | NP_001070962.1 | |
NM_001261403.2 | 1320 | Silent Mutation | CCC,CCT | P,P 349 | NP_001248332.1 | |
NM_001288724.1 | 1320 | Silent Mutation | CCC,CCT | P,P 349 | NP_001275653.1 | |
NM_001322934.1 | 1320 | Silent Mutation | CCC,CCT | P,P 349 | NP_001309863.1 | |
NM_001322935.1 | 1320 | Intron | NP_001309864.1 | |||
NM_002502.5 | 1320 | Silent Mutation | CCC,CCT | P,P 349 | NP_002493.3 | |
XM_011539830.2 | 1320 | Silent Mutation | CCC,CCT | P,P 204 | XP_011538132.1 | |
XM_011539831.2 | 1320 | Silent Mutation | CCC,CCT | P,P 204 | XP_011538133.1 | |
XM_017016278.1 | 1320 | Silent Mutation | CCC,CCT | P,P 530 | XP_016871767.1 |
PSD - pleckstrin and Sec7 domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |