Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTAGCAGTGGGATTTGTGCTTCT[G/T]ATGTTACCCAGGTGAGGTACTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601761 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CASP7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CASP7 - caspase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001227.4 | 79 | Intron | NP_001218.1 | |||
NM_001267056.1 | 79 | Intron | NP_001253985.1 | |||
NM_001267057.1 | 79 | Intron | NP_001253986.1 | |||
NM_001267058.1 | 79 | Intron | NP_001253987.1 | |||
NM_001320911.1 | 79 | Intron | NP_001307840.1 | |||
NM_033338.5 | 79 | Missense Mutation | GAT,TAT | D,Y 30 | NP_203124.1 | |
NM_033339.4 | 79 | UTR 5 | NP_203125.1 | |||
NM_033340.3 | 79 | Intron | NP_203126.1 | |||
XM_006718017.3 | 79 | UTR 5 | XP_006718080.1 | |||
XM_011540260.1 | 79 | Intron | XP_011538562.1 | |||
XM_017016763.1 | 79 | Intron | XP_016872252.1 | |||
XM_017016764.1 | 79 | Intron | XP_016872253.1 |