Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGCAGCGCGCTGCGCCCCAGC[A/C]CCAGCCGCAGCCTCTACGCCTCGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 193060 | ||||||||||||||||||||
Literature Links: |
VIM PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VIM - vimentin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003380.3 | 1630 | Missense Mutation | ACC,CCC | T,P 48 | NP_003371.2 | |
XM_006717500.1 | 1630 | Missense Mutation | ACC,CCC | T,P 48 | XP_006717563.1 |
VIM-AS1 - VIM antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |