Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACTGGTTCGGCTTGTTTTTCAC[C/T]GTCAATGTATTTTTTCCTGCATCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611171 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ENO4 - enolase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242699.1 | 1722 | Intron | NP_001229628.1 | |||
XM_005269815.3 | 1722 | Intron | XP_005269872.1 | |||
XM_006717835.3 | 1722 | Intron | XP_006717898.1 | |||
XM_006717836.3 | 1722 | Intron | XP_006717899.1 | |||
XM_011539797.2 | 1722 | Intron | XP_011538099.1 | |||
XM_011539799.2 | 1722 | Intron | XP_011538101.1 | |||
XM_017016241.1 | 1722 | Intron | XP_016871730.1 |
SHTN1 - shootin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127211.2 | 1722 | Missense Mutation | AGT,GGT | S,G 572 | NP_001120683.1 | |
NM_001258298.1 | 1722 | Missense Mutation | AGT,GGT | S,G 512 | NP_001245227.1 | |
NM_001258299.1 | 1722 | Intron | NP_001245228.1 | |||
NM_001258300.1 | 1722 | Intron | NP_001245229.1 | |||
NM_018330.6 | 1722 | UTR 3 | NP_060800.2 | |||
XM_005270002.3 | 1722 | Intron | XP_005270059.1 | |||
XM_006717931.3 | 1722 | Intron | XP_006717994.1 | |||
XM_017016462.1 | 1722 | Missense Mutation | AGT,GGT | S,G 512 | XP_016871951.1 |