Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAACTTCAGGAAAAACAGTGGCTCC[C/T]CCTGCAGACACATCACTCATCTATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 176710 | ||||||||||||||||||||
Literature Links: |
P4HA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
P4HA1 - prolyl 4-hydroxylase subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000917.3 | 1576 | Silent Mutation | GGA,GGG | G,G 463 | NP_000908.2 | |
NM_001017962.2 | 1576 | Silent Mutation | GGA,GGG | G,G 463 | NP_001017962.1 | |
NM_001142595.1 | 1576 | Silent Mutation | GGA,GGG | G,G 463 | NP_001136067.1 | |
NM_001142596.1 | 1576 | Silent Mutation | GGA,GGG | G,G 445 | NP_001136068.1 |