Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGTGCAGGAGGAGGGGGGCACCCC[A/C]GAGCAGGGGGTTCACAGGGCCCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600827 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDE6C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDE6C - phosphodiesterase 6C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006204.3 | 366 | Silent Mutation | CCA,CCC | P,P 76 | NP_006195.3 | |
XM_017016322.1 | 366 | Intron | XP_016871811.1 |