Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGGTGTGTTGGGCCACGAACAG[C/G]CAGGCCAAAGCCACCACCATGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM24A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM24A - family with sequence similarity 24 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029888.1 | 327 | Missense Mutation | AGC,AGG | S,R 66 | NP_001025059.1 | |
XM_017015638.1 | 327 | Missense Mutation | AGC,AGG | S,R 66 | XP_016871127.1 | |
XM_017015639.1 | 327 | Missense Mutation | AGC,AGG | S,R 66 | XP_016871128.1 |