Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACTCAACCCGGTAACCCCAAA[C/T]GTGCACGGTCCGGCCGGGGCGCGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612105 MIM: 601728 | ||||||||||||||||||||
Literature Links: |
KLLN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLLN - killin, p53-regulated DNA replication inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001126049.1 | 999 | Missense Mutation | ATT,GTT | I,V 17 | NP_001119521.1 |
PTEN - phosphatase and tensin homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000314.6 | 999 | Intron | NP_000305.3 | |||
NM_001304717.2 | 999 | Intron | NP_001291646.2 | |||
NM_001304718.1 | 999 | Intron | NP_001291647.1 |