Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCCCCAACCGCCGCCGCCCCCC[C/G]CACCCGCTCCCAAGGAGTCCCCGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600647 MIM: 613380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMX2 - H6 family homeobox 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HMX3 - H6 family homeobox 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105574.1 | 67 | Missense Mutation | CCA,GCA | P,A 23 | NP_001099044.1 |