Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGAGCGGGATCCCGCGGCTGGGG[C/G]CGCGCCCTTTCGCTTCGTTGCGCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TUBGCP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TUBGCP2 - tubulin gamma complex associated protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256617.1 | 147 | UTR 5 | NP_001243546.1 | |||
NM_001256618.1 | 147 | UTR 5 | NP_001243547.1 | |||
NM_006659.3 | 147 | UTR 5 | NP_006650.1 | |||
XM_017015595.1 | 147 | Intron | XP_016871084.1 | |||
XM_017015596.1 | 147 | Intron | XP_016871085.1 |
ZNF511 - zinc finger protein 511 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145806.3 | 147 | Missense Mutation | GCC,GGC | A,G 31 | NP_665805.2 |