Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAATCAAAACTGGCACACATTCT[A/G]TATCCTTCACAGCACTGCAGGGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616644 | ||||||||||||||||||||
Literature Links: |
CUZD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CUZD1 - CUB and zona pellucida like domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM24B - family with sequence similarity 24 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204364.1 | 526 | Silent Mutation | TAC,TAT | Y,Y 74 | NP_001191293.1 | |
NM_152644.2 | 526 | Silent Mutation | TAC,TAT | Y,Y 74 | NP_689857.2 |
FAM24B-CUZD1 - FAM24B-CUZD1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |