Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGCCCCCCTAGGGTGCTGTGGGA[A/G]CTCCGGCTCCTTGCTTCCTCTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602267 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM8 - ADAM metallopeptidase domain 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109.4 | 2726 | Missense Mutation | GCT,GTT | A,V 820 | NP_001100.3 | |
NM_001164489.1 | 2726 | UTR 3 | NP_001157961.1 | |||
NM_001164490.1 | 2726 | Missense Mutation | GCT,GTT | A,V 729 | NP_001157962.1 | |
XM_011539117.2 | 2726 | Missense Mutation | GCT,GTT | A,V 859 | XP_011537419.2 | |
XM_017015465.1 | 2726 | Missense Mutation | GCT,GTT | A,V 885 | XP_016870954.1 | |
XM_017015466.1 | 2726 | Intron | XP_016870955.1 |