Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAAATGACATTTCTCCTAAAACC[A/G]AAAGTTTGAGAAAGAAAAAGGAGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606357 MIM: 610373 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX21 - DEAD-box helicase 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256910.1 | 499 | Missense Mutation | AAA,GAA | K,E 25 | NP_001243839.1 | |
NM_004728.3 | 499 | Missense Mutation | AAA,GAA | K,E 93 | NP_004719.2 | |
XM_011540336.1 | 499 | Missense Mutation | AAA,GAA | K,E 93 | XP_011538638.1 | |
XM_017016910.1 | 499 | Missense Mutation | AAA,GAA | K,E 25 | XP_016872399.1 |
DDX50 - DEAD-box helicase 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |