Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTGTTGTCCTTTCTTACAGAA[C/T]GTACCTGAAACATGGGAGCCCAGTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM45A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM45A - family with sequence similarity 45 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303111.1 | Intron | NP_001290040.1 | ||||
NM_001303112.1 | Intron | NP_001290041.1 | ||||
NM_001303113.1 | Intron | NP_001290042.1 | ||||
NM_207009.3 | Intron | NP_996892.1 | ||||
XM_006717849.3 | Intron | XP_006717912.1 | ||||
XM_006717850.3 | Intron | XP_006717913.1 | ||||
XM_006717851.3 | Intron | XP_006717914.1 | ||||
XM_006717852.3 | Intron | XP_006717915.1 | ||||
XM_017016261.1 | Intron | XP_016871750.1 | ||||
XM_017016262.1 | Intron | XP_016871751.1 | ||||
XM_017016263.1 | Intron | XP_016871752.1 | ||||
XM_017016264.1 | Intron | XP_016871753.1 | ||||
XM_017016265.1 | Intron | XP_016871754.1 | ||||
XM_017016266.1 | Intron | XP_016871755.1 | ||||
XM_017016267.1 | Intron | XP_016871756.1 |