Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGATCTCCCTCCGGGCGGGCACC[A/G]CAGTGCCACTGGTCCCTGTGAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf82 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf82 - chromosome 10 open reading frame 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HSPA12A - heat shock protein family A (Hsp70) member 12A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025015.2 | 1731 | Missense Mutation | GCG,GTG | A,V 635 | NP_079291.2 | |
XM_005269672.3 | 1731 | Missense Mutation | GCG,GTG | A,V 652 | XP_005269729.1 | |
XM_005269673.4 | 1731 | Missense Mutation | GCG,GTG | A,V 651 | XP_005269730.1 | |
XM_011539579.2 | 1731 | Missense Mutation | GCG,GTG | A,V 651 | XP_011537881.1 | |
XM_011539580.2 | 1731 | Missense Mutation | GCG,GTG | A,V 552 | XP_011537882.1 | |
XM_017016032.1 | 1731 | Missense Mutation | GCG,GTG | A,V 459 | XP_016871521.1 |
LOC105378499 - uncharacterized LOC105378499 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |