Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCAGTGCACCCGGTGCCGCCGC[A/C]GCACGCCCTGGCCGCCCACCCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600035 MIM: 607637 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMX2 - empty spiracles homeobox 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001165924.1 | 1095 | Missense Mutation | CAG,CCG | Q,P 91 | NP_001159396.1 | |
NM_004098.3 | 1095 | Missense Mutation | CAG,CCG | Q,P 91 | NP_004089.1 |
EMX2OS - EMX2 opposite strand/antisense RNA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |