Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGAGGGAAGAAGGACAGGGACT[C/T]GTGTGGCAGGAAGAACTCAGAGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605677 | ||||||||||||||||||||
Literature Links: |
ACSL5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSL5 - acyl-CoA synthetase long-chain family member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016234.3 | 177 | Missense Mutation | TCG,TTG | S,L 22 | NP_057318.2 | |
NM_203379.1 | 177 | Intron | NP_976313.1 | |||
NM_203380.1 | 177 | Intron | NP_976314.1 | |||
XM_017016324.1 | 177 | Missense Mutation | TCG,TTG | S,L 22 | XP_016871813.1 |