Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCGCGCGGGGAGCGCCCAGGATG[C/T]CGCGCGGGGACTCGGAGCAGGTGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613140 | ||||||||||||||||||||
Literature Links: |
TSPAN15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TSPAN15 - tetraspanin 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012339.3 | 129 | Missense Mutation | CCG,TCG | P,S 2 | NP_036471.1 | |
XM_005269666.4 | 129 | Intron | XP_005269723.1 | |||
XM_005269667.3 | 129 | Missense Mutation | CCG,TCG | P,S 2 | XP_005269724.1 | |
XM_011539562.2 | 129 | Intron | XP_011537864.1 | |||
XM_011539563.2 | 129 | Intron | XP_011537865.1 | |||
XM_017016010.1 | 129 | Missense Mutation | CCG,TCG | P,S 2 | XP_016871499.1 |