Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCCCCAGCCAGATGATGAAAC[A/T]GATACCTCCTATTTTGAAGCCAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616618 MIM: 608221 | ||||||||||||||||||||
Literature Links: |
ACBD5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MASTL - microtubule associated serine/threonine kinase like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001172303.2 | 3416 | Silent Mutation | ACA,ACT | T,T 858 | NP_001165774.1 | |
NM_001172304.2 | 3416 | Silent Mutation | ACA,ACT | T,T 819 | NP_001165775.1 | |
NM_001320756.1 | 3416 | Silent Mutation | ACA,ACT | T,T 862 | NP_001307685.1 | |
NM_001320757.1 | 3416 | Silent Mutation | ACA,ACT | T,T 863 | NP_001307686.1 | |
NM_032844.4 | 3416 | Silent Mutation | ACA,ACT | T,T 857 | NP_116233.2 | |
XM_005252631.3 | 3416 | Silent Mutation | ACA,ACT | T,T 820 | XP_005252688.1 | |
XM_005252632.3 | 3416 | Silent Mutation | ACA,ACT | T,T 786 | XP_005252689.1 | |
XM_006717519.3 | 3416 | Silent Mutation | ACA,ACT | T,T 829 | XP_006717582.1 | |
XM_006717520.3 | 3416 | Intron | XP_006717583.1 | |||
XM_011519751.2 | 3416 | Silent Mutation | ACA,ACT | T,T 526 | XP_011518053.1 | |
XM_017016852.1 | 3416 | Silent Mutation | ACA,ACT | T,T 823 | XP_016872341.1 | |
XM_017016853.1 | 3416 | Silent Mutation | ACA,ACT | T,T 814 | XP_016872342.1 | |
XM_017016854.1 | 3416 | Intron | XP_016872343.1 | |||
XM_017016855.1 | 3416 | Intron | XP_016872344.1 | |||
XM_017016856.1 | 3416 | UTR 3 | XP_016872345.1 |