Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATGTTTGCGGTCCACTTGATGGC[A/G]TTTTACTTCAGCAAGCTGAAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HKDC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HKDC1 - hexokinase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025130.3 | 157 | Silent Mutation | GCA,GCG | A,A 8 | NP_079406.3 | |
XM_011540195.2 | 157 | Silent Mutation | GCA,GCG | A,A 8 | XP_011538497.1 |
LOC101928994 - uncharacterized LOC101928994 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |