Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAATGCGTATTTTGGGTTGCGTG[A/C]AAGAGGTGAATGAACTGGAACTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612333 MIM: 615204 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1307 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR1307 - microRNA 1307 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDCD11 - programmed cell death 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014976.1 | 398 | Missense Mutation | AAA,CAA | K,Q 91 | NP_055791.1 | |
XM_005269647.3 | 398 | Missense Mutation | AAA,CAA | K,Q 91 | XP_005269704.1 | |
XM_011539538.2 | 398 | Missense Mutation | AAA,CAA | K,Q 91 | XP_011537840.1 | |
XM_011539539.2 | 398 | Missense Mutation | AAA,CAA | K,Q 91 | XP_011537841.1 | |
XM_011539540.1 | 398 | Missense Mutation | AAA,CAA | K,Q 8 | XP_011537842.1 |
USMG5 - up-regulated during skeletal muscle growth 5 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |