Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCACCAGCCGCTTGGTGTCCTTGC[A/G]CTTCAGGGGGCCCGGCTTGAGCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604158 MIM: 610243 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFRP5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFRP5 - secreted frizzled related protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003015.3 | 875 | Missense Mutation | CGC,TGC | R,C 237 | NP_003006.2 |
ZFYVE27 - zinc finger FYVE-type containing 27 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |