Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAACTGTGACTGTCTAAATGGAGG[A/G]ACATGTGTGTCCAACAAGTACTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 191840 | ||||||||||||||||||||
Literature Links: |
C10orf55 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C10orf55 - chromosome 10 open reading frame 55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001791.2 | 346 | Missense Mutation | TCC,TTC | S,F 2 | NP_001001791.2 |
PLAU - plasminogen activator, urokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145031.2 | 346 | Silent Mutation | GGA,GGG | G,G 20 | NP_001138503.1 | |
NM_001319191.1 | 346 | UTR 5 | NP_001306120.1 | |||
NM_002658.4 | 346 | Silent Mutation | GGA,GGG | G,G 37 | NP_002649.1 | |
XM_011539866.2 | 346 | Silent Mutation | GGA,GGG | G,G 37 | XP_011538168.1 |