Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAATCCGAGGAATCTTCTGCAC[A/C]GGTGAGTCCTCCGCGGAGGGGGACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EDRF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EDRF1 - erythroid differentiation regulatory factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202438.1 | 400 | Missense Mutation | CAG,CCG | Q,P 36 | NP_001189367.1 | |
NM_015608.2 | 400 | Missense Mutation | CAG,CCG | Q,P 36 | NP_056423.2 | |
XM_011539622.1 | 400 | Intron | XP_011537924.1 | |||
XM_017016066.1 | 400 | UTR 5 | XP_016871555.1 |
FLJ37035 - uncharacterized LOC399821 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |