Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTCACACAATCCCAGTGCCACAT[A/T]TGTCACACATAGGCAACTTCTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605900 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSM6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACSM6 - acyl-CoA synthetase medium-chain family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDLIM1 - PDZ and LIM domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020992.3 | 724 | Intron | NP_066272.1 | |||
XM_011540330.1 | 724 | Missense Mutation | AAA,ATA | K,I 159 | XP_011538632.1 |