Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCGGCGACCTGAGCCGCGGAGA[C/G]GTGCCGGAACCTCTGTGTGCCTGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609208 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KAZALD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KAZALD1 - Kazal type serine peptidase inhibitor domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319303.1 | 1285 | UTR 5 | NP_001306232.1 | |||
NM_030929.4 | 1285 | Missense Mutation | GAC,GAG | D,E 120 | NP_112191.2 | |
XM_005270194.3 | 1285 | Missense Mutation | GAC,GAG | D,E 120 | XP_005270251.1 | |
XM_017016714.1 | 1285 | Missense Mutation | GAC,GAG | D,E 120 | XP_016872203.1 | |
XM_017016715.1 | 1285 | Missense Mutation | GAC,GAG | D,E 120 | XP_016872204.1 |