Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTCCTCCTCAGCCTCTTTCTG[A/G]TGCTTGAGCTTCTGGTGCTTTTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRP12 - ribosomal RNA processing 12 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145114.1 | 3730 | Silent Mutation | CAC,CAT | H,H 1136 | NP_001138586.1 | |
NM_001284337.1 | 3730 | Silent Mutation | CAC,CAT | H,H 1097 | NP_001271266.1 | |
NM_015179.3 | 3730 | Silent Mutation | CAC,CAT | H,H 1197 | NP_055994.2 | |
XM_011539555.1 | 3730 | Silent Mutation | CAC,CAT | H,H 1169 | XP_011537857.1 | |
XM_011539556.2 | 3730 | Intron | XP_011537858.1 |