Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGCCTGCCCTCCGTGAGCTACG[C/G]CAGCTCCGGGGCCAGCGTGAGCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602292 MIM: 615853 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECHS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECHS1 - enoyl-CoA hydratase, short chain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3944 - microRNA 3944 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PAOX - polyamine oxidase (exo-N4-amino) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152911.3 | 427 | Missense Mutation | GCC,GGC | A,G 116 | NP_690875.1 | |
NM_207127.2 | 427 | Missense Mutation | GCC,GGC | A,G 116 | NP_997010.1 | |
NM_207128.2 | 427 | Missense Mutation | GCC,GGC | A,G 116 | NP_997011.1 |