Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTTGTCAGCCAGGGCTGAACCCC[C/T]GCAGGTAAGGGGGAGGGGGCGTCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615570 MIM: 611129 | ||||||||||||||||||||
Literature Links: |
SFXN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SFXN2 - sideroflexin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WBP1L - WW domain binding protein 1-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083913.1 | 170 | Missense Mutation | CCG,CTG | P,L 29 | NP_001077382.1 | |
NM_017787.4 | 170 | Intron | NP_060257.4 | |||
XM_011539913.2 | 170 | Intron | XP_011538215.1 | |||
XM_017016360.1 | 170 | UTR 5 | XP_016871849.1 |